Gene Transcripts

Gene Symbol
Il13ra1
Entrez Gene ID
16164
Gene Name
interleukin 13 receptor, alpha 1
Gene Aliases
AI882074, CD213a1, IL-13R-alpha-1, IL-13r[a], Il13ra, NR4
Chromosome Location
Chr. X: 36112139 - 36171262 on Build GRCm38
UniGene ID
Mm.24208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK080147.1 - 5 584
AK082889.1 - 5 579
AK089984.1 - 5 615
AK143292.1 - 5 584
AK154675.1 - 5 639
BC021472.1 - 1 38
BC052425.1 - 5 607 6835500
BC059939.1 - 5 575 6477224
S80963.1 - 5 612

View Full Product Details