Gene Transcripts

Gene Symbol
Itih2
Entrez Gene ID
16425
Gene Name
inter-alpha trypsin inhibitor, heavy chain 2
Gene Aliases
AI747202, Intin2, Itih-2
Chromosome Location
Chr. 2: 10094591 - 10130683 on Build GRCm38
UniGene ID
Mm.182043
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010582.3 NP_034712.2 12 1487
GenBank mRNA AK133782.1 - 12 1386
BC034341.1 - 12 1461 4211576
X70392.1 - 12 1473

View Full Product Details