Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025922.2 | NP_080198.2 | 2 | 112 | |
GenBank mRNA | AB100501.1 | - | 2 | 124 | |
AK008279.1 | - | 2 | 152 | ||
AK009248.1 | - | 2 | 125 | ||
AK138101.1 | - | 2 | 152 | ||
AK139770.1 | - | 2 | 110 | ||
AK153806.1 | - | 2 | 152 | ||
AK153904.1 | - | 2 | 131 | ||
BC026508.1 | - | 2 | 112 | 4512398 | |
BC094466.1 | - | 2 | 100 | 4936811 |