Gene Transcripts

Gene Symbol
HOXD8
Entrez Gene ID
3234
Gene Name
homeobox D8
Gene Aliases
HOX4, HOX4E, HOX5.4
Chromosome Location
Chr. 2: 176129694 - 176132695 on Build GRCh38
UniGene ID
Hs.301963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199746.1 NP_001186675.1 2 1244
NM_001199747.1 NP_001186676.1 3 272
NM_019558.3 NP_062458.1 2 1247
GenBank mRNA BC038709.1 - 2 1094
BC090853.1 - 3 943 6180525
BQ429720.1 - 3 222

View Full Product Details