Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205043.1 | NP_001191972.1 | 4 | 1062 | |
NM_001205044.1 | NP_001191973.1 | 5 | 735 | ||
NM_021878.3 | NP_068678.1 | 5 | 781 | ||
GenBank mRNA | AK044888.1 | - | 4 | 966 | |
AK045214.1 | - | 4 | 867 | ||
AK147226.1 | - | 4 | 1062 | ||
AK164134.1 | - | 3 | 386 | ||
BC026056.1 | - | 4 | 969 | ||
BC050129.1 | - | 4 | 983 | ||
BC051010.1 | - | 4 | 965 | ||
BC052444.1 | - | 5 | 652 | 6406875 | |
BC060695.1 | - | 5 | 652 | 6406875 | |
D31967.1 | - | 5 | 671 |