Gene Transcripts

Gene Symbol
Kcnab3
Entrez Gene ID
16499
Gene Name
potassium voltage-gated channel, shaker-related subfamily, beta member 3
Gene Aliases
C330022D06Rik, Kcnab4, mKv(beta)4
Chromosome Location
Chr. 11: 69326258 - 69333041 on Build GRCm38
UniGene ID
Mm.232472
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010599.4 NP_034729.3 4 793
GenBank mRNA AK029750.1 - 3 382
AK031806.1 - 4 774
AK044102.1 - 4 748
AK046194.1 - 4 783
AK049316.1 - 4 794
AK083134.1 - 4 795
BC017518.1 - 4 719 4507029

View Full Product Details