Gene Transcripts

Gene Symbol
Krt19
Entrez Gene ID
16669
Gene Name
keratin 19
Gene Aliases
AI663979, EndoC, K19, Krt-1.19, Krt1-19
Chromosome Location
Chr. 11: 100140810 - 100145919 on Build GRCm38
UniGene ID
Mm.439699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008471.2 NP_032497.1 5 944
GenBank mRNA AB472075.1 - 5 944
BC034561.1 - 5 937 2655016
BC085304.1 - 5 944 6437375
M28698.1 - 5 939

View Full Product Details