Gene Transcripts
- Gene Symbol
- Lmo4
- Entrez Gene ID
- 16911
- Gene Name
- LIM domain only 4
- Gene Aliases
- A730077C12Rik, Crp3, Etohi4
- Chromosome Location
- Chr. 3: 144188530 - 144205255 on Build GRCm38
- UniGene ID
- Mm.29187
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161769.1 |
NP_001155241.1
|
2, 3 |
382 |
|
NM_001161770.1 |
NP_001155242.1
|
1, 2 |
514 |
|
NM_010723.3 |
NP_034853.1
|
2, 3 |
669 |
|
GenBank mRNA |
AF074600.1 |
- |
1, 2 |
230 |
|
AF096996.1 |
- |
2, 3 |
398 |
|
AF102817.1 |
- |
2, 3 |
299 |
|
AK080534.1 |
- |
2, 3 |
317 |
|
AK138278.1 |
- |
1, 2 |
515 |
|
AK159590.1 |
- |
2, 3 |
596 |
|
BC003488.1 |
- |
1, 2 |
381 |
2811636 |
BC004661.1 |
- |
2, 3 |
322 |
|
BC010278.1 |
- |
2, 3 |
412 |
3589532 |
BC100382.1 |
- |
2, 3 |
274 |
6487478 |
BY267947.1 |
- |
2 |
376 |
|
View Full Product Details