Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161769.1 | NP_001155241.1 | 4 | 531 | |
NM_001161770.1 | NP_001155242.1 | 3 | 663 | ||
NM_010723.3 | NP_034853.1 | 4 | 818 | ||
GenBank mRNA | AF074600.1 | - | 3 | 379 | |
AF096996.1 | - | 4 | 547 | ||
AF102817.1 | - | 4 | 448 | ||
AK080534.1 | - | 4 | 466 | ||
AK138278.1 | - | 3 | 664 | ||
AK159590.1 | - | 4 | 745 | ||
BC003488.1 | - | 3 | 530 | 2811636 | |
BC004661.1 | - | 4 | 471 | ||
BC010278.1 | - | 4 | 561 | 3589532 | |
BC100382.1 | - | 4 | 423 | 6487478 |