Gene Transcripts

Gene Symbol
Lyst
Entrez Gene ID
17101
Gene Name
lysosomal trafficking regulator
Gene Aliases
D13Sfk13, beige, bg
Chromosome Location
Chr. 13: 13590339 - 13778803 on Build GRCm38
UniGene ID
Mm.342337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010748.2 NP_034878.2 8, 9 3870
GenBank mRNA AK089635.1 - 6, 7 3631
L77884.1 - 8, 9 3870
U70015.1 - 8, 9 3870

View Full Product Details