Gene Transcripts

Gene Symbol
Mcm5
Entrez Gene ID
17218
Gene Name
minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae)
Gene Aliases
AA617332, AI324988, AL033333, Cdc46, Mcmd5, P1-CDC46
Chromosome Location
Chr. 8: 75109509 - 75128439 on Build GRCm38
UniGene ID
Mm.5048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK033196.1 - 4 473
AK051795.1 - 4 443
AK133826.1 - 4 474
BC090645.1 - 4 490
BC094416.1 - 4 438 5707724
CF948662.1 - 4 490
D26090.1 - 3 496

View Full Product Details