Gene Transcripts

Gene Symbol
Melk
Entrez Gene ID
17279
Gene Name
maternal embryonic leucine zipper kinase
Gene Aliases
AI327312, MPK38, mKIAA0175
Chromosome Location
Chr. 4: 44300876 - 44364675 on Build GRCm38
UniGene ID
Mm.268668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010790.2 NP_034920.2 10, 11 1026
GenBank mRNA AK010498.1 - 10, 11 992
AK011932.1 - 10, 11 1026
AK129076.1 - 10, 11 979
AK137000.1 - 10, 11 1052
AK145021.1 - 10, 11 961
AK145316.1 - 10, 11 935
AK161029.1 - 10, 11 1035
AK164138.1 - 10, 11 993
AK167457.1 - 10, 11 950
BC020416.1 - 9, 10 861
BC085276.1 - 10, 11 1059 30443236
L76158.1 - 10, 11 957
X95351.1 - 10, 11 964

View Full Product Details