Gene Transcripts
- Gene Symbol
- HSPA7
- Entrez Gene ID
- 3311
- Gene Name
- heat shock protein family A (Hsp70) member 7
- Gene Aliases
- HSP70B
- Chromosome Location
- Chr. 1: 161606059 - 161608551 on Build GRCh38
- UniGene ID
- Hs.662180
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024151.1 |
|
1 |
1156 |
|
GenBank mRNA |
CA391777.1 |
- |
1 |
467 |
|
- Gene Symbol
- HSPA6
- Entrez Gene ID
- 3310
- Gene Name
- heat shock protein family A (Hsp70) member 6
- Gene Aliases
- HSP70B'
- Chromosome Location
- Chr. 1: 161524540 - 161526897 on Build GRCh38
- UniGene ID
- Hs.654614
- Species
- Homo sapiens
View Full Product Details