Gene Transcripts

Gene Symbol
Morc1
Entrez Gene ID
17450
Gene Name
microrchidia 1
Gene Aliases
Morc
Chromosome Location
Chr. 16: 48431237 - 48630905 on Build GRCm38
UniGene ID
Mm.250060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010816.1 NP_034946.1 19 2022
GenBank mRNA AF084945.1 - 19 2022
BC030893.1 - 19 1971 4487929

View Full Product Details