Gene Transcripts

Gene Symbol
Mrc1
Entrez Gene ID
17533
Gene Name
mannose receptor, C type 1
Gene Aliases
AW259686, CD206, MR
Chromosome Location
Chr. 2: 14229414 - 14332023 on Build GRCm38
UniGene ID
Mm.2019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008625.2 NP_032651.2 28 4139
GenBank mRNA AK079897.1 - 1 91
BC141338.1 - 28 4091 9055950
Z11974.1 - 28 4139

View Full Product Details