Gene Transcripts
- Gene Symbol
- Msi1
- Entrez Gene ID
- 17690
- Gene Name
- musashi RNA-binding protein 1
- Gene Aliases
- Msi1h, Musahi1, m-Msi-1
- Chromosome Location
- Chr. 5: 115359083 - 115455698 on Build GRCm38
- UniGene ID
- Mm.391995
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008629.1 |
NP_032655.1
|
11, 12 |
853 |
|
GenBank mRNA |
D49654.1 |
- |
11, 12 |
853 |
|
View Full Product Details