Gene Transcripts

Gene Symbol
HSPG2
Entrez Gene ID
3339
Gene Name
heparan sulfate proteoglycan 2
Gene Aliases
HSPG, PLC, PRCAN, SJA, SJS, SJS1
Chromosome Location
Chr. 1: 21822232 - 21937297 on Build GRCh38
UniGene ID
Hs.562227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291860.1 NP_001278789.1 37 4771
NM_005529.6 NP_005520.4 37 4768
XM_011541318.2 XP_011539620.1 42 5252
XM_017001120.1 XP_016856609.1 38 4898
XM_017001121.1 XP_016856610.1 37 4847
XM_017001122.1 XP_016856611.1 37 4844
GenBank mRNA M85289.1 - 37 4768
X62515.1 - 37 4734

View Full Product Details