Gene Transcripts

Gene Symbol
Mut
Entrez Gene ID
17850
Gene Name
methylmalonyl-Coenzyme A mutase
Gene Aliases
D230010K02Rik, Mcm
Chromosome Location
Chr. 17: 40934685 - 40961989 on Build GRCm38
UniGene ID
Mm.259884
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008650.3 NP_032676.2 3 815
GenBank mRNA AK146309.1 - 3 775
BC019175.1 - 3 797 5007694
X51941.1 - 2 726

View Full Product Details