Gene Transcripts

Gene Symbol
Mvk
Entrez Gene ID
17855
Gene Name
mevalonate kinase
Gene Aliases
2310010A05Rik, AI256848, AI414037, MK
Chromosome Location
Chr. 5: 114444257 - 114460591 on Build GRCm38
UniGene ID
Mm.28088
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF137598.1 - 3 259
AK009261.1 - 5 369
AK031084.1 - 3 1040
AK149630.1 - 4 356
AK181570.1 - 2 113
AK187523.1 - 2 113
AK187805.1 - 2 149
AK210419.1 - 11
BC005606.1 - 4 312 3498140

View Full Product Details