Gene Transcripts
- Gene Symbol
- Myl6
- Entrez Gene ID
- 17904
- Gene Name
- myosin, light polypeptide 6, alkali, smooth muscle and non-muscle
- Gene Aliases
- ESMLC, LC17, LC17-GI, LC17-NM, LC17A, LC17B, MLC-3, MLC1SM, MLC3SM, MLC3nm, Myln
- Chromosome Location
- Chr. 10: 128490861 - 128493896 on Build GRCm38
- UniGene ID
- Mm.337074
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010860.3 |
NP_034990.1
|
3, 4 |
198 |
|
GenBank mRNA |
AK143885.1 |
- |
3, 4 |
228 |
|
AK167106.1 |
- |
3, 4 |
228 |
|
AK167191.1 |
- |
3, 4 |
234 |
|
AK167859.1 |
- |
3, 4 |
228 |
|
AK168450.1 |
- |
3, 4 |
249 |
|
AK168869.1 |
- |
3, 4 |
230 |
|
BC026760.1 |
- |
3, 4 |
198 |
3990090 |
BC081470.1 |
- |
3, 4 |
215 |
6824167 |
BC089485.1 |
- |
3, 4 |
221 |
30277988 |
BC092214.1 |
- |
3, 4 |
206 |
5684607 |
View Full Product Details