Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008670.2 | NP_032696.2 | 11 | 3022 | |
GenBank mRNA | AF007769.1 | - | 10 | 3119 | |
AF135491.1 | - | 11 | 3022 | ||
BC132413.1 | - | 10 | 3069 | 40130690 | |
BC145251.1 | - | 10 | 2339 |