Gene Transcripts

Gene Symbol
HTR4
Entrez Gene ID
3360
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
5-HT4, 5-HT4R
Chromosome Location
Chr. 5: 148451032 - 148654527 on Build GRCh38
UniGene ID
Hs.483773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000870.6 NP_000861.1 6 1354
NM_001040169.2 NP_001035259.1 5 888
NM_001040172.2 NP_001035262.2 5 888
NM_001040173.2 NP_001035263.1 5 888
NM_001286410.1 NP_001273339.1 5 888
NM_199453.3 NP_955525.1 5 888
NR_104445.1 6 1354
GenBank mRNA AJ011371.1 - 5 843
AJ131724.1 - 6 981
AJ131725.1 - 5 847
AJ131726.1 - 5 847
AJ278979.1 - 5 847
AJ278980.1 - 5 847
AJ278981.1 - 5 847
AJ278982.1 - 5 847
AJ519673.1 - 5 860
AJ633645.1 - 5 841
AK308471.1 - 6 1354
AM712912.1 - 5 841
BC074755.2 - 5 882 30915214
BC095497.1 - 5 847 6971934
Y08756.1 - 5 859
Y09586.1 - 5 843
Y10437.1 - 5 841
Y12505.1 - 5 859
Y12506.1 - 5 859
Y12507.1 - 5 859
Y13584.1 - 5 841

View Full Product Details