Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008721.4 | NP_032747.2 | 6 | 918 | |
GenBank mRNA | AK146466.1 | - | 6 | 807 | |
BC003320.1 | - | 6 | 885 | 3155595 | |
BC023887.1 | - | 5 | 952 | 5151868 | |
L03814.1 | - | 6 | 722 | ||
X67209.1 | - | 6 | 722 |