Gene Transcripts

Gene Symbol
Olfr19
Entrez Gene ID
18316
Gene Name
olfactory receptor 19
Gene Aliases
M12, MOR140-1, MTPCR15
Chromosome Location
Chr. 16: 16673050 - 16673979 on Build GRCm38
UniGene ID
Mm.222834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146335.1 NP_666447.1 1 559
GenBank mRNA BC132306.1 - 1 597
X89683.1 - 1 185

View Full Product Details