Gene Transcripts

Gene Symbol
ALDOA
Entrez Gene ID
226
Gene Name
aldolase, fructose-bisphosphate A
Gene Aliases
ALDA, GSD12, HEL-S-87p
Chromosome Location
Chr. 16: 30053090 - 30070420 on Build GRCh38
UniGene ID
Hs.513490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000034.3 NP_000025.1 14 2159
NM_001127617.2 NP_001121089.1 9 1381
NM_001243177.1 NP_001230106.1 10 1502
NM_184041.2 NP_908930.1 9 1348
NM_184043.2 NP_908932.1 9 1320
XM_011545768.2 XP_011544070.1 11 2022
GenBank mRNA AK026577.1 - 9 1197
AK098778.1 - 12 2200
AK301993.1 - 10 1314
BC000367.2 - 6 760
BC004333.1 - 14 2079 3630974
BC010660.1 - 9 1266 3869603
BC012880.1 - 9 1194 3459009
BC013614.1 - 9 1277 3880857
BC015888.2 - 9 1190 3911623
BC016170.1 - 6 760
BC016800.1 - 10 1218 4072487
BU855133.1 - 4 467
BX647566.1 - 9 1170
CR536528.1 - 8 1072
CR541880.1 - 8 1072
FJ474908.1 - 10 1239
M11560.1 - 10 1239
X05236.1 - 9 1218

View Full Product Details