Gene Transcripts

Gene Symbol
Olfr47
Entrez Gene ID
18346
Gene Name
olfactory receptor 47
Gene Aliases
IB12, MOR261-9
Chromosome Location
Chr. 6: 43235610 - 43236575 on Build GRCm38
UniGene ID
Mm.222840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146370.1 NP_666482.1 1 243
GenBank mRNA AF102521.1 - 1 57
BC120658.1 - 1 275 40129581
BC120660.1 - 1 275 40129583

View Full Product Details