Gene Transcripts

Gene Symbol
Pabpc2
Entrez Gene ID
18459
Gene Name
poly(A) binding protein, cytoplasmic 2
Gene Aliases
Pabp, Pabp+, Pabp2
Chromosome Location
Chr. 18: 39773497 - 39776082 on Build GRCm38
UniGene ID
Mm.9493
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011033.2 NP_035163.1 1 1929
GenBank mRNA AK077154.1 - 1 1929
BC051134.1 - 1 1909 6704229
CN837699.1 - 1 89
X75959.1 - 1 1905

View Full Product Details