Gene Transcripts

Gene Symbol
ARPC4
Entrez Gene ID
10093
Gene Name
actin related protein 2/3 complex subunit 4
Gene Aliases
ARC20, P20-ARC
Chromosome Location
Chr. 3: 9792495 - 9807105 on Build GRCh38
UniGene ID
Hs.323342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024959.2 NP_001020130.1 6 1075
NM_001024960.2 NP_001020131.1 6 930
NM_001198780.1 NP_001185709.1 6 929
NM_005718.4 NP_005709.1 6 1441
GenBank mRNA BC012596.2 - 5 860

View Full Product Details