Gene Transcripts
- Gene Symbol
- IFRD1
- Entrez Gene ID
- 3475
- Gene Name
- interferon related developmental regulator 1
- Gene Aliases
- PC4, TIS7
- Chromosome Location
- Chr. 7: 112423144 - 112477203 on Build GRCh38
- UniGene ID
- Hs.744747
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007245.2 |
NP_001007246.1
|
4, 5 |
754 |
|
NM_001197079.1 |
NP_001184008.1
|
3, 4 |
644 |
|
NM_001197080.1 |
NP_001184009.1
|
3, 4 |
479 |
|
NM_001550.3 |
NP_001541.2
|
3, 4 |
545 |
|
NR_120333.1 |
|
3, 4 |
465 |
|
XM_011516142.2 |
XP_011514444.1
|
3, 4 |
540 |
|
XM_011516143.2 |
XP_011514445.1
|
3, 4 |
725 |
|
XM_017012151.1 |
XP_016867640.1
|
3, 4 |
792 |
|
GenBank mRNA |
AK225222.1 |
- |
3, 4 |
518 |
|
AK297509.1 |
- |
3, 4 |
290 |
|
AK298894.1 |
- |
3, 4 |
285 |
|
AK308066.1 |
- |
4, 5 |
729 |
|
AK313211.1 |
- |
4, 5 |
754 |
|
AK315967.1 |
- |
3, 4 |
477 |
|
AU280361.1 |
- |
3, 4 |
542 |
|
BC001272.1 |
- |
3, 4 |
498 |
3458309 |
BT019889.1 |
- |
3, 4 |
284 |
|
BT019890.1 |
- |
3, 4 |
284 |
|
View Full Product Details