Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163530.1 | NP_001157002.1 | 5 | 584 | |
NM_011164.2 | NP_035294.2 | 5 | 587 | ||
GenBank mRNA | AK017520.1 | - | 5 | 580 | |
AK017521.1 | - | 5 | 581 | ||
AK017547.1 | - | 5 | 585 | ||
AK017562.1 | - | 5 | 572 | ||
AK017579.1 | - | 5 | 581 | ||
AK017584.1 | - | 5 | 578 | ||
AK077493.1 | - | 5 | 584 | ||
AK133645.1 | - | 5 | 578 | ||
AK133736.1 | - | 5 | 581 | ||
AK133752.1 | - | 5 | 581 | ||
AK161553.1 | - | 5 | 577 | ||
AK161557.1 | - | 5 | 581 | ||
AK161565.1 | - | 5 | 581 | ||
AK161594.1 | - | 5 | 578 | ||
AK161619.1 | - | 5 | 585 | ||
BC061141.1 | - | 5 | 580 | 30250937 | |
X02892.1 | - | 5 | 579 | ||
X04418.1 | - | 5 | 581 |