Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011172.2 | NP_035302.2 | 4 | 665 | |
GenBank mRNA | AF120279.1 | - | 4 | 622 | |
AK144068.1 | - | 4 | 635 | ||
BC008548.1 | - | 4 | 489 | ||
BC125327.1 | - | 4 | 468 | 40129842 | |
U80020.1 | - | 4 | 614 |