Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008964.4 | NP_032990.1 | 1 | 1080 | |
GenBank mRNA | AK036474.1 | - | 1 | 525 | |
AK036531.1 | - | 1 | 999 | ||
AK054132.1 | - | 1 | 601 | ||
AK086904.1 | - | 1 | 994 | ||
AK136577.1 | - | 1 | 93 | ||
AK137515.1 | - | 1 | 290 | ||
BC005440.1 | - | 1 | 588 | 3487087 | |
D50589.1 | - | 1 | 371 |