Gene Transcripts
- Gene Symbol
- Ptma
- Entrez Gene ID
- 19231
- Gene Name
- prothymosin alpha
- Gene Aliases
- Thym
- Chromosome Location
- Chr. 1: 86526736 - 86530698 on Build GRCm38
- UniGene ID
- Mm.19187
- Species
- Mus musculus
- Gene Symbol
- Gm12504
- Entrez Gene ID
- 623796
- Gene Name
- prothymosin alpha pseudogene
- Gene Aliases
- OTTMUSG00000007209
- Chromosome Location
- Chr. 4: 44121893 - 44124123 on Build GRCm38
- UniGene ID
- Mm.490409
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038173.1 |
- |
1 |
2154 |
|
View Full Product Details