Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291019.1 | NP_001277948.1 | 4 | 1473 | |
NM_001291020.1 | NP_001277949.1 | 5 | 1275 | ||
NM_007547.4 | NP_031573.2 | 4 | 1473 | ||
GenBank mRNA | AB018194.1 | - | 4 | 1039 | |
AF072543.1 | - | 4 | 1029 | ||
AF332079.1 | - | 4 | 1039 | ||
AF332080.1 | - | 4 | 1039 | ||
AK159617.1 | - | 5 | 1282 | ||
AK195466.1 | - | 1 | 38 | ||
BC062197.1 | - | 4 | 1064 | 5368250 | |
D85785.1 | - | 4 | 1323 | ||
D87967.1 | - | 4 | 1438 | ||
D87968.1 | - | 4 | 1438 | ||
U89694.1 | - | 4 | 1050 | ||
Y10349.1 | - | 4 | 1084 |