Gene Transcripts
- Gene Symbol
- Rab18
- Entrez Gene ID
- 19330
- Gene Name
- RAB18, member RAS oncogene family
- Gene Aliases
- AA959686
- Chromosome Location
- Chr. 18: 6765014 - 6791606 on Build GRCm38
- UniGene ID
- Mm.132802
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278447.1 |
NP_001265376.1
|
4, 5 |
372 |
|
NM_181070.6 |
NP_851415.1
|
4, 5 |
354 |
|
GenBank mRNA |
AB232612.1 |
- |
4, 5 |
256 |
|
AK045514.1 |
- |
4, 5 |
317 |
|
AK140901.1 |
- |
4, 5 |
322 |
|
AK143673.1 |
- |
4, 5 |
330 |
|
AK146177.1 |
- |
4, 5 |
279 |
|
AK146397.1 |
- |
4, 5 |
279 |
|
AK154920.1 |
- |
4, 5 |
331 |
|
AK216482.1 |
- |
1, 2 |
64 |
|
BC056351.1 |
- |
4, 5 |
308 |
6807284 |
L04966.1 |
- |
4, 5 |
260 |
|
X80333.1 |
- |
4, 5 |
256 |
|
View Full Product Details