Gene Transcripts

Gene Symbol
IL1B
Entrez Gene ID
3553
Gene Name
interleukin 1 beta
Gene Aliases
IL-1, IL1-BETA, IL1F2
Chromosome Location
Chr. 2: 112829758 - 112836842 on Build GRCh38
UniGene ID
Hs.126256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000576.2 NP_000567.1 5 444
XM_017003988.1 XP_016859477.1 4 445
GenBank mRNA AK313983.1 - 4 1164
BC008678.1 - 5 444 3875593
BT007213.1 - 4 357
CR407679.1 - 4 357
K02770.1 - 5 443
M15330.1 - 5 443
M54933.1 - 5 444
X02532.1 - 5 434
X56087.1 - 5 414

View Full Product Details