Gene Transcripts

Gene Symbol
Rbp3
Entrez Gene ID
19661
Gene Name
retinol binding protein 3, interstitial
Gene Aliases
Irbp, Rbp-3
Chromosome Location
Chr. 14: 33954022 - 33964216 on Build GRCm38
UniGene ID
Mm.436622
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015745.2 NP_056560.1 3 3357
GenBank mRNA BC017610.1 - 3 3357
BC023069.1 - 3 648
BC034121.1 - 3 476
BC035273.1 - 3 1789
BC040282.1 - 3 3341
BC042480.1 - 3 3319 5363924
BC042509.1 - 3 550
BC083133.1 - 3 3376 6492729

View Full Product Details