Gene Transcripts

Gene Symbol
Bex1
Entrez Gene ID
19716
Gene Name
brain expressed gene 1
Gene Aliases
AA407839, AU018692, Bex4, Rex3
Chromosome Location
Chr. X: 136213972 - 136215513 on Build GRCm38
UniGene ID
Mm.422943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009052.2 NP_033078.2 3 742
GenBank mRNA AF051347.1 - 3 675
AF097438.1 - 3 687
AK003429.1 - 1 357
BC058805.1 - 1 1014
BC061179.1 - 3 687 6531808
BC089464.1 - 3 736 6594252
CB194115.1 - 3 664

View Full Product Details