Gene Transcripts

Gene Symbol
Rit1
Entrez Gene ID
19769
Gene Name
Ras-like without CAAX 1
Gene Aliases
RIBB, ROC1, Rit
Chromosome Location
Chr. 3: 88716840 - 88731048 on Build GRCm38
UniGene ID
Mm.4009
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163310.1 NP_001156782.1 3 188
NM_009069.4 NP_033095.1 4 333
GenBank mRNA AK005357.1 - 4 294
AK014126.1 - 4 324
AK018785.1 - 4 281
AK164196.1 - 4 324
AK168254.1 - 4 292
BC012694.1 - 4 293 4023732
BY284832.1 - 4 336
CV562852.1 - 4 301
CX242151.1 - 3 136
U71205.1 - 4 270

View Full Product Details