Gene Transcripts
- Gene Symbol
- Rpl26
- Entrez Gene ID
- 19941
- Gene Name
- ribosomal protein L26
- Gene Aliases
- SIG-20
- Chromosome Location
- Chr. 11: 68901566 - 68904537 on Build GRCm38
- UniGene ID
- Mm.306548
- Species
- Mus musculus
- Gene Symbol
- Ceacam1
- Entrez Gene ID
- 26365
- Gene Name
- carcinoembryonic antigen-related cell adhesion molecule 1
- Gene Aliases
- Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1, bb-1, mCEA1, mmCGM1, mmCGM1a, mmCGM2
- Chromosome Location
- Chr. 7: 25461702 - 25477625 on Build GRCm38
- UniGene ID
- Mm.322502
- Species
- Mus musculus
- Gene Symbol
- Gm15772
- Entrez Gene ID
- 100034726
- Gene Name
- ribosomal protein L26 pseudogene
- Gene Aliases
- ENSMUSG00000062353, OTTMUSG00000026108
- Chromosome Location
- Chr. 5: 3236390 - 3236928 on Build GRCm38
- UniGene ID
- Mm.3229
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK146116.1 |
- |
1 |
371 |
|
X80699.1 |
- |
1 |
378 |
|
View Full Product Details