Gene Transcripts

Gene Symbol
Rpl9
Entrez Gene ID
20005
Gene Name
ribosomal protein L9
Gene Aliases
Chromosome Location
Chr. 5: 65388364 - 65391431 on Build GRCm38
UniGene ID
Mm.300271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011292.2 NP_035422.1 2, 3 104
GenBank mRNA AF260271.1 - 1, 2 74
AK012331.1 - 2, 3 74
AK012445.1 - 2, 3 74
AK017388.1 - 2, 3 74
AK017427.1 - 2, 3 74
AK053431.1 - 1, 2 457
AK084278.1 - 2, 3 75
AK088166.1 - 2, 3 74
AK153562.1 - 2, 3 74
AK159980.1 - 2, 3 74
AK168326.1 - 2, 3 72
AK169217.1 - 2, 3 72
BC013165.1 - 1, 2 64 2655358
BC081435.1 - 1, 2 50 6824107
BC083166.1 - 1, 2 55 6397429
BC083329.1 - 1, 2 53 6824346
BC089319.1 - 1, 2 53 6823996
BC093526.1 - 1, 2 53 5150493
BY072215.1 - 2, 3 106

View Full Product Details