Gene Transcripts

Gene Symbol
IL7
Entrez Gene ID
3574
Gene Name
interleukin 7
Gene Aliases
IL-7
Chromosome Location
Chr. 8: 78675870 - 78805523 on Build GRCh38
UniGene ID
Hs.591873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000880.3 NP_000871.1 3 815
NM_001199886.1 NP_001186815.1 3 815
NM_001199887.1 NP_001186816.1 3 815
NM_001199888.1 NP_001186817.1 3 815
XM_011517522.2 XP_011515824.1 3 755
XM_011517523.2 XP_011515825.1 3 755
XM_017013397.1 XP_016868886.1 3 699
GenBank mRNA AB102879.1 - 2 214
AB102883.1 - 1 61
AB102893.1 - 2 214
AK226000.1 - 4 959
AK291538.1 - 3 815
BC047698.1 - 3 803 5748841
CD013923.1 - 3 220
GQ919059.1 - 2 139
J04156.1 - 3 598

View Full Product Details