Gene Transcripts
- Gene Symbol
- Ryr1
- Entrez Gene ID
- 20190
- Gene Name
- ryanodine receptor 1, skeletal muscle
- Gene Aliases
- AI528790, RYR-1, Ryr, skrr
- Chromosome Location
- Chr. 7: 29003340 - 29125179 on Build GRCm38
- UniGene ID
- Mm.439745
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009109.2 |
NP_033135.2
|
68, 69 |
10491 |
|
GenBank mRNA |
AY268935.1 |
- |
68, 69 |
10491 |
|
View Full Product Details