Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011314.2 | NP_035444.1 | 3 | 182 | |
GenBank mRNA | BC024606.1 | - | 2 | 156 | |
BC034858.1 | - | 1 | 1342 | ||
L22190.1 | - | 2 | 180 | ||
M11130.1 | - | 3 | 182 | ||
U60438.1 | - | 2 | 146 |