Gene Transcripts

Gene Symbol
IL13RA1
Entrez Gene ID
3597
Gene Name
interleukin 13 receptor subunit alpha 1
Gene Aliases
CD213A1, CT19, IL-13Ra, NR4
Chromosome Location
Chr. X: 118726954 - 118794533 on Build GRCh38
UniGene ID
Hs.496646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001560.2 NP_001551.1 9 1135
XM_011531336.1 XP_011529638.1 9 1413
XM_011531337.1 XP_011529639.1 2 1442
XM_017029507.1 XP_016884996.1 8 1152
GenBank mRNA AB209849.1 - 9 1090
AK123515.1 - 2 1098
AK296958.1 - 4 364
AK313467.1 - 9 1124
BC009960.2 - 9 1122 4300487
BC015768.1 - 9 1139 4868206
U81379.3 - 9 1132
Y09328.1 - 9 1125
Y10659.1 - 9 1135

View Full Product Details