Gene Transcripts
- Gene Symbol
- Slc31a2
- Entrez Gene ID
- 20530
- Gene Name
- solute carrier family 31, member 2
- Gene Aliases
- AI604396, CTR2
- Chromosome Location
- Chr. 4: 62280393 - 62298412 on Build GRCm38
- UniGene ID
- Mm.292539
- Species
- Mus musculus
- Gene Symbol
- Aldh9a1
- Entrez Gene ID
- 56752
- Gene Name
- aldehyde dehydrogenase 9, subfamily A1
- Gene Aliases
- AA139417, Abaldh, ESTM40, TMABA-DH, Tmabadh
- Chromosome Location
- Chr. 1: 167349991 - 167368530 on Build GRCm38
- UniGene ID
- Mm.292539
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK177761.1 |
- |
|
180 |
|
View Full Product Details