Gene Transcripts

Gene Symbol
Snapin
Entrez Gene ID
20615
Gene Name
SNAP-associated protein
Gene Aliases
25kDa, AA407989, AV026596, Bloc1s7, Snap25bp, Snapap
Chromosome Location
Chr. 3: 90488026 - 90491013 on Build GRCm38
UniGene ID
Mm.331182
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133854.3 NP_598615.1 2 201
GenBank mRNA AF086838.1 - 2 162
AK007458.1 - 2 182
AK035147.1 - 2 198
AK048007.1 - 2 190
AK050151.1 - 2 202
AK077840.1 - 2 192
AK080718.1 - 2 192
AK086994.1 - 1 133
AK135006.1 - 2 193
AK152057.1 - 2 170
BC048691.1 - 2 173 6511395

View Full Product Details