Gene Transcripts

Gene Symbol
Sntb2
Entrez Gene ID
20650
Gene Name
syntrophin, basic 2
Gene Aliases
Snt2
Chromosome Location
Chr. 8: 106935750 - 107014192 on Build GRCm38
UniGene ID
Mm.30228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009229.4 NP_033255.1 3 829
GenBank mRNA AK040423.1 - 3 830
AK080059.1 - 3 826
AK131687.1 - 3 854
AK165009.1 - 3 831
AK165900.1 - 3 806
BC138850.1 - 3 828 8861872
BC145442.1 - 3 828 9054000
U00678.1 - 3 828

View Full Product Details