Gene Transcripts

Gene Symbol
Ssr4
Entrez Gene ID
20832
Gene Name
signal sequence receptor, delta
Gene Aliases
AW743063, C78062, Trap
Chromosome Location
Chr. X: 73787028 - 73790828 on Build GRCm38
UniGene ID
Mm.831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166480.1 NP_001159952.1 6 782
NM_009279.3 NP_033305.1 6 664
GenBank mRNA AK007921.1 - 6 620
AK013723.1 - 6 626
AK160089.1 - 6 630
AK187990.1 - 2 199
AK207479.1 - 2 199
AK214704.1 - 2 199
BC012971.1 - 3 2559
BQ747160.1 - 6 567
X90582.1 - 6 745

View Full Product Details