Gene Transcripts

Gene Symbol
Sult1c1
Entrez Gene ID
20888
Gene Name
sulfotransferase family, cytosolic, 1C, member 1
Gene Aliases
(PST)G, P-SULT, ST1C1, Stp2, Sult1a2, mOLFST
Chromosome Location
Chr. 17: 53961615 - 53990631 on Build GRCm38
UniGene ID
Mm.10108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018751.2 NP_061221.2 6 732
GenBank mRNA AF033653.1 - 6 743
BC045149.1 - 6 732 6308175

View Full Product Details